4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate
072148-1G
072148-250MG
102523-348EA
212.31
USD
102523-348
102523-350
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester ≥95%
4,6-Dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O₂ MW: 209.25 g/mol |
MDL Number:
MFCD08061257 CAS Number: 657428-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...