7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥95%
Supplier: Matrix Scientific
Synonyms:
7-(4-Bromobutoxy)-1,2,3,4-tetrahydro-2-oxoquinoline
, 7-(4-Bromobutoxy)-3,4-dihydroquinolin-2(1H)-one
072127-1G
072127-10G
072127-5G
102523-238EA
346.15
USD
102523-238
102523-236
102523-234
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone ≥95%
7-(4-Bromobutoxy)-3,4-dihydro-2(1H)-quinolinone
Hazard: Irritant
Formula:
C₁₃H₁₆BrNO₂ MW: 298.18 g/mol |
MDL Number:
MFCD06658540 CAS Number: 129722-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...