2-Bromo-3-nitrotoluene ≥95%

Supplier: Matrix Scientific

072105-5G
102523-118EA 32.31 USD
102523-118
2-Bromo-3-nitrotoluene ≥95%
2-Bromo-3-nitrotoluene

Hazard: Irritant-Harmful/Light Sensitive

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 135…136 °C (8 mmHg)
Melting Pt: 38…41 °C
MDL Number: MFCD00134555
CAS Number: 41085-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR