2,6-Dibromopyrazine ≥95%

Supplier: Matrix Scientific

072094-5G
102523-064EA 113.85 USD
102523-064
2,6-Dibromopyrazine ≥95%
2,6-Dibromopyrazine

Hazard: Irritant

Formula: C₄H₂Br₂N₂
MW: 237.882 g/mol
MDL Number: MFCD08061601
CAS Number: 23229-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR