2-(2-Bromophenyl)oxazole ≥95%

Supplier: Matrix Scientific

072087-5G 072087-1G 072087-250MG
102523-026EA 3435.38 USD
102523-026 102523-024 102523-022
2-(2-Bromophenyl)oxazole ≥95%
2-(2-Bromophenyl)oxazole

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD08234434
CAS Number: 92346-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR