2-Methylisonicotinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Methylpyridine-4-carboxylic acid
072070-5G
072070-1G
102522-936EA
167.69
USD
102522-936
102522-934
2-Methylisonicotinic acid ≥95%
2-Methylisonicotinic acid
Hazard: Irritant
Formula:
C₇H₇NO₂ MW: 137.14 Melting Pt: 295…300 °C |
MDL Number:
MFCD00513435 CAS Number: 4021-11-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...