4-Chlorosalicylaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-2-hydroxybenzaldehyde
072067-5G
102522-920EA
120
USD
102522-920
4-Chlorosalicylaldehyde ≥95%
4-Chlorosalicylaldehyde
Hazard: Irritant
Formula:
C₇H₅ClO₂ MW: 156.5664 g/mol |
MDL Number:
MFCD06252499 CAS Number: 2420-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...