1-Boc-4-(4-benzylpiperazin-1-yl)-4-methylpiperidine ≥95%

Supplier: Matrix Scientific

072063-1G 072063-5G
102522-898EA 533.85 USD
102522-898 102522-900
1-Boc-4-(4-benzylpiperazin-1-yl)-4-methylpiperidine ≥95%
1-Boc-4-(4-benzylpiperazin-1-yl)-4-methylpiperidine

Hazard: Irritant

Formula: C₂₂H₃₅N₃O₂
MDL Number: MFCD09743718
CAS Number: 1189489-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR