2-(4-Fluorophenyl)thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

072057-1G 072057-5G
102522-872EA 710.77 USD
102522-872 102522-874
2-(4-Fluorophenyl)thiazole-4-carboxylic acid ≥95%
2-(4-Fluorophenyl)thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆FNO₂S
MW: 223.23 g/mol
MDL Number: MFCD07339052
CAS Number: 863668-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR