2-Amino-5-bromo-4-dimethoxymethylpyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-4-(dimethoxymethyl)pyrimidin-2-amine

072017-1G 072017-5G
102522-652EA 710.77 USD
102522-652 102522-654
2-Amino-5-bromo-4-dimethoxymethylpyrimidine ≥95%
2-Amino-5-bromo-4-dimethoxymethylpyrimidine

Hazard: Irritant

Formula: C₇H₁₀BrN₃O₂
MW: 248.09 g/mol
MDL Number: MFCD08460359
CAS Number: 914347-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR