4,6-Dichloroquinazoline ≥95%

Supplier: Matrix Scientific

072010-5G 072010-1G
102522-616EA 116.92 USD
102522-616 102522-618
4,6-Dichloroquinazoline ≥95%
4,6-Dichloroquinazoline

Hazard: Irritant

Formula: C₈H₄Cl₂N₂
MW: 199.0368 g/mol
MDL Number: MFCD00089492
CAS Number: 7253-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR