2-[4-(6-Amino-2-methylpyrimidin-4-yl)piperazin-1-yl]ethanol ≥95%

Supplier: Matrix Scientific

072007-5G 072007-1G 072007-250MG
102522-598EA 355.38 USD
102522-598 102522-600 102522-602
2-[4-(6-Amino-2-methylpyrimidin-4-yl)piperazin-1-yl]ethanol ≥95%
2-[4-(6-Amino-2-methylpyrimidin-4-yl)piperazin-1-yl]ethanol

Hazard: Irritant

Formula: C₁₁H₁₉N₅O
MW: 237.31 g/mol
MDL Number: MFCD08275728
CAS Number: 914347-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR