(3-Bromoisoxazol-5-yl)methylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Bromoisoxazol-5-yl)methanamine hydrochloride

071992-1G 071992-5G
102522-512EA 201.54 USD
102522-512 102522-514
(3-Bromoisoxazol-5-yl)methylamine hydrochloride ≥95%
(3-Bromoisoxazol-5-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₄H₆BrClN₂O
MW: 213.47 g/mol
MDL Number: MFCD08705903
CAS Number: 90802-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR