1-Boc-4-(5-bromopyrimidin-2-yl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(5-bromopyrimidin-2-yl)piperazine-1-carboxylate

071989-5G
102522-496EA 130.77 USD
102522-496
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine ≥95%
1-Boc-4-(5-bromopyrimidin-2-yl)piperazine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₄O₂
MW: 343.23 g/mol
MDL Number: MFCD07375175
CAS Number: 374930-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR