1-Bromo-3-chloro-5-nitrobenzene ≥95%
Supplier: Matrix Scientific
071987-10G
071987-1G
071987-5G
102522-482EA
61.54
USD
102522-482
102522-484
102522-486
1-Bromo-3-chloro-5-nitrobenzene ≥95%
1-Bromo-3-chloro-5-nitrobenzene
Hazard: Irritant
Formula:
C₆H₃BrClNO₂ MW: 236.4500 g/mol |
MDL Number:
MFCD08275708 CAS Number: 219817-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...