5-Bromo-8-nitroisoquinoline ≥95%

Supplier: Matrix Scientific

071979-5G
102522-438EA 144.62 USD
102522-438
5-Bromo-8-nitroisoquinoline ≥95%
5-Bromo-8-nitroisoquinoline

Hazard: Irritant

Formula: C₉H₅BrN₂O₂
MW: 253.0500 g/mol
MDL Number: MFCD02091227
CAS Number: 63927-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR