4-(Boc-amino)cyclohexanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Aminocyclohexan-1-one, N-BOC protectedtert-Butyl (4-oxocyclohexyl)carbamate

071959-5G
102522-320EA 73.85 USD
102522-320
4-(Boc-amino)cyclohexanone ≥95%
4-(Boc-amino)cyclohexanone

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.2700 g/mol
MDL Number: MFCD00798168
CAS Number: 179321-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR