2-Amino-3-chloro-5-nitropyridine ≥95%

Supplier: Matrix Scientific

071957-5G 071957-1G 071957-10G
102522-308EA 1076.92 USD
102522-308 102522-310 102522-306
2-Amino-3-chloro-5-nitropyridine ≥95%
2-Amino-3-chloro-5-nitropyridine

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.5600 g/mol
MDL Number: MFCD00160299
CAS Number: 22353-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR