2-Bromopyrazine ≥95%

Supplier: Matrix Scientific

071944-5G 071944-1G
102522-234EA 294.23 USD
102522-234 102522-232
2-Bromopyrazine ≥95%
2-Bromopyrazine

Purity: 95+%
CAS: 56423-63-3
MDL Number: MFCD08275680
Molecular Formula: C4H3BrN2
Molecular Weight: 158,99
Hazard: Irritant/Refrigerate

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR