Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate
071934-1G
071934-5G
071934-10G
102522-182EA
692.31
USD
102522-182
102522-178
102522-180
Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate ≥95%
Methyl-5-bromo-2-((tert-butoxycarbonyl)amino)thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂O₄S MW: 337.1900 g/mol |
MDL Number:
MFCD07782035 CAS Number: 914349-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...