tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-(4-chlorophenyl)piperazine-1-carboxylate

071933-250MG 071933-1G
102522-174EA 710.77 USD
102522-174 102522-176
tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-2-(4-chlorophenyl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O₂
MW: 296.7924 g/mol
MDL Number: MFCD08275679
CAS Number: 769944-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR