(S)-(+)-1-Boc-3-hydroxypyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate(S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

071932-100G
102522-172EA 157.55 USD
102522-172
(S)-(+)-1-Boc-3-hydroxypyrrolidine ≥95%
(S)-(+)-1-Boc-3-hydroxypyrrolidine

Hazard: Irritant

Formula: C₉H₁₇NO₃
MW: 187.2362 g/mol
MDL Number: MFCD01317839
CAS Number: 101469-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR