4-Ethyl-6-hydroxy-2-methylpyrimidine ≥95%

Supplier: Matrix Scientific

071929-1G 071929-5G 071929-10G
102522-150EA 355.38 USD
102522-150 102522-152 102522-154
4-Ethyl-6-hydroxy-2-methylpyrimidine ≥95%
4-Ethyl-6-hydroxy-2-methylpyrimidine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD03265397
CAS Number: 52421-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR