2-Amino-3,5-dibromopyrazine ≥98%

Supplier: Matrix Scientific

071924-25G
102833-510EA 212.75 USD
102833-510
2-Amino-3,5-dibromopyrazine ≥98%
2-Amino-3,5-dibromopyrazine

Hazard: Toxic

Formula: C₄H₃Br₂N₃
MW: 252.90 g/mol
Melting Pt: 114…117 °C
MDL Number: MFCD00673150
CAS Number: 24241-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR