5-Ethoxy-1H-indole ≥95%

Supplier: Matrix Scientific

071921-5G 071921-10G 071921-1G
102522-110EA 1787.69 USD
102522-110 102522-106 102522-108
5-Ethoxy-1H-indole ≥95%
5-Ethoxy-1H-indole

Hazard: Irritant

Formula: C₁₀H₁₁NO
MW: 161.2004 g/mol
MDL Number: MFCD07364533
CAS Number: 10501-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR