5-Nitrophthalazine ≥95%

Supplier: Matrix Scientific

071916-1G 071916-5G
102522-084EA 1690.77 USD
102522-084 102522-082
5-Nitrophthalazine ≥95%
5-Nitrophthalazine

Hazard: Irritant

Formula: C₈H₅N₃O₂
MW: 175.1400 g/mol
MDL Number: MFCD08236796
CAS Number: 89898-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR