2-Amino-4-nitrobenzamide ≥95%

Supplier: Matrix Scientific

071912-1G 071912-5G
102522-058EA 100 USD
102522-058 102522-060
2-Amino-4-nitrobenzamide ≥95%
2-Amino-4-nitrobenzamide

Hazard: Irritant

Formula: C₇H₇N₃O₃
MW: 181.1500 g/mol
MDL Number: MFCD07368613
CAS Number: 31930-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR