tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Boc-4-(4-aminophenyl)piperazine
, tert-Butyl 4-(4-aminophenyl)piperazine-1-carboxylate
071903-10G
071903-5G
071903-1G
102522-010EA
249.23
USD
102522-010
102522-012
102522-008
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(4-aminophenyl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₅H₂₃N₃O₂ MW: 277.36 g/mol |
MDL Number:
MFCD04115065 CAS Number: 170911-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...