1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrophenyl)piperazine ≥95%
Supplier: Matrix Scientific
071901-5G
071901-10G
071901-1G
102522-000EA
963.08
USD
102522-000
102521-996
102521-998
1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrophenyl)piperazine ≥95%
1-(Bis(4-fluorophenyl)methyl)-4-(4-nitrophenyl)piperazine
Hazard: Irritant
Formula:
C₂₃H₂₁F₂N₃O₂ |
CAS Number:
851472-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...