4-(4-Nitrophenyl)morpholine ≥95%

Supplier: Matrix Scientific

071899-10G 071899-5G
102521-986EA 355.38 USD
102521-986 102521-988
4-(4-Nitrophenyl)morpholine ≥95%
4-(4-Nitrophenyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃
MDL Number: MFCD00023317
CAS Number: 10389-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR