2-Methyl-1-(4-nitrobenzyl)-1H-imidazole ≥95%

Supplier: Matrix Scientific

071896-5G 071896-10G 071896-1G
102521-970EA 463.08 USD
102521-970 102521-972 102521-968
2-Methyl-1-(4-nitrobenzyl)-1H-imidazole ≥95%
2-Methyl-1-(4-nitrobenzyl)-1H-imidazole

Hazard: Irritant

Formula: C₁₁H₁₁N₃O₂
CAS Number: 56643-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR