1-(4-Nitrobenzyl)-1H-imidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Nitrobenzyl)-1H-imidazole(Chunks or pellets)

071892-1G 071892-5G 071892-10G
102521-948EA 549.23 USD
102521-948 102521-946 102521-944
1-(4-Nitrobenzyl)-1H-imidazole ≥95%
1-(4-Nitrobenzyl)-1H-imidazole

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MW: 203.1974 g/mol
MDL Number: MFCD01799096
CAS Number: 18994-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR