1-(4-Nitrobenzyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

071890-1G 071890-5G 071890-10G
102521-936EA 963.08 USD
102521-936 102521-934 102521-932
1-(4-Nitrobenzyl)-1H-pyrazole ≥95%
1-(4-Nitrobenzyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
CAS Number: 110525-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR