3-Thiazol-2-ylbenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Thiazol-2-yl-benzaldehyde

071871-10G 071871-5G 071871-1G
102521-828EA 1787.69 USD
102521-828 102521-826 102521-824
3-Thiazol-2-ylbenzaldehyde ≥95%
3-Thiazol-2-ylbenzaldehyde

Hazard: Irritant

Formula: C₁₀H₇NOS
MW: 189.24 g/mol
MDL Number: MFCD06797781
CAS Number: 885465-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR