Pseudoisocytidine ≥95%

Supplier: Matrix Scientific

071867-250MG 071867-1G
102521-804EA 4146.15 USD
102521-804 102521-802
Pseudoisocytidine ≥95%
Pseudoisocytidine

Hazard: Irritant

Formula: C₉H₁₃N₃O₅
CAS Number: 57100-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR