4-(4-Ethylpiperazin-1-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

071856-10G 071856-5G 071856-1G
102521-738EA 769.23 USD
102521-738 102521-740 102521-736
4-(4-Ethylpiperazin-1-yl)benzoic acid ≥95%
4-(4-Ethylpiperazin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.2900 g/mol
MDL Number: MFCD05864834
CAS Number: 784130-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR