4-(2-Aminopyrimidin-5-yl)benzoic acid ≥95%
Supplier: Matrix Scientific
071833-5G
071833-1G
071833-10G
102521-600EA
320
USD
102521-600
102521-604
102521-602
4-(2-Aminopyrimidin-5-yl)benzoic acid ≥95%
4-(2-Aminopyrimidin-5-yl)benzoic acid
Hazard: Irritant
Formula:
C₁₁H₉N₃O₂ MW: 215.2100 g/mol |
MDL Number:
MFCD05864814 CAS Number: 222987-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...