3-Pyrimidin-5-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Pyrimidin-5-yl)benzoic acid

071828-1G 071828-10G 071828-5G
102521-570EA 320 USD
102521-570 102521-574 102521-572
3-Pyrimidin-5-ylbenzoic acid ≥95%
3-Pyrimidin-5-ylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
MW: 200.1900 g/mol
MDL Number: MFCD05864810
CAS Number: 852180-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR