4-Pyrid-4-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyridin-4-yl-benzoic acid

071827-5G
102521-566EA 361.54 USD
102521-566
4-Pyrid-4-ylbenzoic acid ≥95%
4-Pyrid-4-ylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₉NO₂
MW: 199.2100 g/mol
MDL Number: MFCD04114574
CAS Number: 4385-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR