4-Pyrid-2-ylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Pyridyl)benzoic acid4-(Pyridin-2-yl)benzoic acid

071826-5G 071826-1G
102521-560EA 390.77 USD
102521-560 102521-558
4-Pyrid-2-ylbenzoic acid ≥95%
4-Pyrid-2-ylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₉NO₂
MW: 199.2100 g/mol
MDL Number: MFCD01830093
CAS Number: 4385-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR