5-(4-Fluorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
071816-1G
071816-10G
071816-5G
102521-498EA
533.85
USD
102521-498
102521-502
102521-500
5-(4-Fluorophenyl)-1H-pyrazole-3-carboxylic acid ≥95%
5-(4-Fluorophenyl)-1H-pyrazole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₇FN₂O₂ MW: 206.1700 g/mol |
MDL Number:
MFCD04073723 CAS Number: 870704-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...