a-Tosyl(4-fluorobenzyl)isocyanide ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-Fluorophenyl)(isocyano)methyl 4-methylphenyl sulphone
, a-Tosyl-(4-fluorobenzyl) isocyanide
071803-1G
071803-5G
102521-424EA
1076.92
USD
102521-424
102521-422
a-Tosyl(4-fluorobenzyl)isocyanide ≥95%
a-Tosyl(4-fluorobenzyl)isocyanide
Hazard: Irritant
Formula:
C₁₅H₁₂FNO₂S MW: 289.32 g/mol |
MDL Number:
MFCD03787932 CAS Number: 165806-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...