5-(4-Methoxyphenyl)pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

071796-1G 071796-5G
102521-384EA 355.38 USD
102521-384 102521-386
5-(4-Methoxyphenyl)pyrimidin-2-amine ≥95%
5-(4-Methoxyphenyl)pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₁H₁₁N₃O
CAS Number: 31408-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR