1-(4-(Hydroxymethyl)phenyl)piperidin-4-ol ≥95%
Supplier: Matrix Scientific
071784-1G
071784-5G
071784-10G
102521-314EA
284.62
USD
102521-314
102521-318
102521-316
1-(4-(Hydroxymethyl)phenyl)piperidin-4-ol ≥95%
1-(4-(Hydroxymethyl)phenyl)piperidin-4-ol
Hazard: Irritant
Formula:
C₁₂H₁₇NO₂ |
CAS Number:
914349-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...