1-(Piperidine-4-carbonyl)piperidin-4-one hydrochloride ≥95%

071780-1G 071780-10G 071780-5G
102521-294EA 784.62 USD
102521-294 102521-292 102521-290
1-(Piperidine-4-carbonyl)piperidin-4-one hydrochloride ≥95%
1-(Piperidine-4-carbonyl)piperidin-4-one hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₉ClN₂O₂
CAS Number: 1189684-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR