(4-Benzylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride ≥95%
Supplier: Matrix Scientific
071779-5G
071779-1G
071779-10G
102521-284EA
284.62
USD
102521-284
102521-288
102521-286
(4-Benzylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride ≥95%
(4-Benzylpiperazin-1-yl)(piperidin-4-yl)methanone dihydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₇Cl₂N₃O |
CAS Number:
1185013-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...