4-Amino-3-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
071752-10G
071752-5G
071752-1G
102521-130EA
1535.38
USD
102521-130
102521-132
102521-128
4-Amino-3-methylbenzenesulfonamide ≥95%
4-Amino-3-methylbenzenesulphonamide
Hazard: Irritant
Formula:
C₇H₁₀N₂O₂S MW: 186.2300 g/mol |
MDL Number:
MFCD00466840 CAS Number: 53297-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...