2-Amino-4-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

071746-1G 071746-5G
102521-098EA 1841.54 USD
102521-098 102521-096
2-Amino-4-nitrobenzonitrile ≥95%
2-Amino-4-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
MDL Number: MFCD04117876
CAS Number: 87376-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR