4-Bromophenylglyoxal hydrate ≥95%
Supplier: Matrix Scientific
071636-5G
071636-10G
071636-1G
102520-474EA
861.54
USD
102520-474
102520-470
102520-472
4-Bromophenylglyoxal hydrate ≥95%
4-Bromophenylglyoxal hydrate
Hazard: Irritant
Formula:
C₈H₇BrO₃ |
MDL Number:
MFCD09260478 CAS Number: 80352-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...