4-Biphenylglyoxal hydrate ≥95%
Supplier: Matrix Scientific
071633-10G
071633-5G
071633-1G
102520-452EA
213.85
USD
102520-452
102520-454
102520-456
4-Biphenylglyoxal hydrate ≥95%
4-Biphenylglyoxal hydrate
Hazard: Irritant
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol |
MDL Number:
MFCD01673220 CAS Number: 1145-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...